Skip to Content
Merck
All Photos(1)

Key Documents

716294

Sigma-Aldrich

2-Fluoro-4-(trifluoromethyl)pyridine

≥94%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$89.50

$89.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$89.50

About This Item

Empirical Formula (Hill Notation):
C6H3F4N
CAS Number:
Molecular Weight:
165.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$89.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥94%

form

liquid

refractive index

n20/D 1.400

density

1.355 g/mL at 25 °C

functional group

fluoro

SMILES string

Fc1cc(ccn1)C(F)(F)F

InChI

1S/C6H3F4N/c7-5-3-4(1-2-11-5)6(8,9)10/h1-3H

InChI key

DFNQBXZKPUBEIX-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of aminopyridines via amination reactions

Catalytic ligand for:
  • Regioselective preparation of tetramethylbiphenyls via aerobic oxidative coupling of xylene catalyzed by palladium

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.0 °F - closed cup

Flash Point(C)

23.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service