Skip to Content
Merck
All Photos(1)

Key Documents

716049

Sigma-Aldrich

4,4′-Bis(mercaptomethyl)biphenyl

97%

Synonym(s):

[1,1′-Biphenyl]-4,4′-dimethanethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14S2
CAS Number:
Molecular Weight:
246.39
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

form

solid

mp

148-152 °C

storage temp.

2-8°C

SMILES string

SCc1ccc(cc1)-c2ccc(CS)cc2

InChI

1S/C14H14S2/c15-9-11-1-5-13(6-2-11)14-7-3-12(10-16)4-8-14/h1-8,15-16H,9-10H2

InChI key

XFHIDPOTWOFDEM-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianli Zhao et al.
The journal of physical chemistry letters, 6(24), 4986-4990 (2015-12-01)
Incorporating single-electron tunneling (SET) of metallic nanoparticles (NPs) into modern electronic devices offers great promise to enable new properties; however, it is technically very challenging due to the necessity to integrate ultrasmall (<10 nm) particles into the devices. The nanosize

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service