Skip to Content
Merck
All Photos(1)

Documents

715034

Sigma-Aldrich

3-(Diphenylphosphino)propionic acid

97%

Synonym(s):

(2-Carboxyethyl)diphenylphosphine, 4,4-Diphenyl-4-phosphabutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15O2P
CAS Number:
Molecular Weight:
258.25
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

130-134 °C

functional group

phosphine

SMILES string

OC(=O)CCP(c1ccccc1)c2ccccc2

InChI

1S/C15H15O2P/c16-15(17)11-12-18(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12H2,(H,16,17)

InChI key

OTSIFUHGOBFOTH-UHFFFAOYSA-N

Application

Reactant for:
  • Organocatalytic asymmetric aziridination of imines and diazo compounds
  • Phosphine-mediated conversion of azides into diazo compounds
  • Preparation of rhodium catalysts for hydroformylation
  • Synthesis of reagents for the Mitsunobu reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dania Ahmed et al.
AMB Express, 7(1), 210-210 (2017-11-23)
Biofilm formation by pathogenic bacteria is one of the major threats in hospital related infections, hence inhibiting and eradicating biofilms has become a primary target for developing new anti-infection approaches. The present study was aimed to develop novel antibiofilm agents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service