Skip to Content
Merck
All Photos(1)

Key Documents

712787

Sigma-Aldrich

2-Azido-β-D-glucose tetraacetate

≥98% (HPLC)

Synonym(s):

1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-β-D-glucopyranose, 2-Azido-2-deoxy-3,4,6-tri-O-acetyl-β-D-glucopyranosyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19N3O9
CAS Number:
Molecular Weight:
373.32
Beilstein:
4337930
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +2.9±0.4°, c = 1 in chloroform

reaction suitability

reaction type: click chemistry

storage temp.

2-8°C

SMILES string

CC(=O)OC[C@H]1O[C@@H](OC(C)=O)C(N=[N+]=[N-])[C@@H](OC(C)=O)[C@@H]1OC(O)=O

InChI

1S/C13H17N3O10/c1-5(17)22-4-8-10(26-13(20)21)11(23-6(2)18)9(15-16-14)12(25-8)24-7(3)19/h8-12H,4H2,1-3H3,(H,20,21)/t8-,9?,10-,11-,12-/m1/s1

InChI key

NKGUHWHPMGOFTK-XBINAETPSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A short synthesis of 1,3,4,6-tetra-O-acetyl-2-azido-2-deoxy-beta-D- glucopyranose and the corresponding alpha-glucosyl chloride from D-mannose.
V Pavliak et al.
Carbohydrate research, 210, 333-337 (1991-03-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service