Skip to Content
Merck
All Photos(1)

Key Documents

711969

Sigma-Aldrich

(R)-3-(Boc-amino)-5-hexenoic acid

98%

Synonym(s):

Boc-(R)-3-amino-5-hexenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
Beilstein:
9397198
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.5% (HPLC)
98%

form

lumps

optical activity

[α]/D -20.0±1°, c = 1 in ethanol

optical purity

enantiomeric excess: ≥99.0%

application(s)

peptide synthesis

functional group

allyl
amine
carboxylic acid

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@H](CC=C)CC(O)=O

InChI

1S/C11H19NO4/c1-5-6-8(7-9(13)14)12-10(15)16-11(2,3)4/h5,8H,1,6-7H2,2-4H3,(H,12,15)(H,13,14)/t8-/m1/s1

InChI key

RFHPQLCVYMBPRF-MRVPVSSYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service