Skip to Content
Merck
All Photos(2)

Key Documents

711144

Sigma-Aldrich

Potassium 1H-pyrazole-3-trifluoroborate

Synonym(s):

Potassium 1H-pyrazole-5-trifluoroborate, Potassium pyrazole-5-trifluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$98.90

$98.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$98.90

About This Item

Empirical Formula (Hill Notation):
C3H3BF3KN2
CAS Number:
Molecular Weight:
173.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$98.90


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

196-200 °C

SMILES string

[K+].F[B-](F)(F)c1cc[nH]n1

InChI

1S/C3H3BF3N2.K/c5-4(6,7)3-1-2-8-9-3;/h1-2H,(H,8,9);/q-1;+1

InChI key

QNHYBMZHGRWURK-UHFFFAOYSA-N

Application

An Organotrifluoroborate involved in Suzuki-Miyaura cross-coupling reactions

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service