Skip to Content
Merck
All Photos(1)

Key Documents

709859

Sigma-Aldrich

TEMPO on silica gel

Synonym(s):

2,2,6,6-Tetramethyl-1-piperinyloxy, free radical on silica gel, SiliaCAT® TEMPO

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$157.00
25 G
$618.00

$157.00


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
5 G
$157.00
25 G
$618.00

About This Item

MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$157.00


Estimated to ship on30 March 2025


Request a Bulk Order

form

solid

Quality Level

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

FeCl3.6H2O/TEMPO (2,2,6,6-tetramethylpiperidine 1-oxyl radical) in the presence of silica gel has been employed as catalyst for oxidation of alcohols to the corresponding aldehydes and ketones.[1] Doping of stable organic nitroxyl radical TEMPO, over different silica matrices affords reusable highly selective catalysts for various alcohol oxidation reactions.[2] TEMPO-supported on silica gel catalyzed-Anelli oxidation of alcohols has been reported.[3]

Application

Recyclable catalyst in the Anelli oxidation of alcohols.

Legal Information

SiliaCat is a registered trademark of Silicycle, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Fey et al.
The Journal of organic chemistry, 66(24), 8154-8159 (2001-11-28)
The application of silica-supported TEMPO as a recyclable catalyst in the Anelli oxidation of alcohols is reported. The catalyst is easily obtained in a one-step reductive amination procedure starting from a commercially available aminopropyl-functionalized silica. Details of the synthesis of
Sol-gel ormosils doped with TEMPO as recyclable catalysts for the selective oxidation of alcohols.
Ciriminna R, et al.
Advanced Synthesis & Catalysis, 344(2), 159-163 (2002)
Selective aerobic oxidation of alcohols catalyzed by iron chloride hexahydrate/TEMPO in the presence of silica gel.
Wang L, et al.
Applied Organometallic Chemistry, 26(1), 37-43 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service