Skip to Content
Merck
All Photos(1)

Key Documents

709700

Sigma-Aldrich

2-(Boc-amino)benzylamine

Synonym(s):

(2-Aminomethyl-phenyl)-carbamic acid tert-butyl ester, tert-Butyl N-(2-aminomethylphenyl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$174.00

$174.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$174.00

About This Item

Empirical Formula (Hill Notation):
C12H18N2O2
CAS Number:
Molecular Weight:
222.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$174.00


Check Cart for Availability

Request a Bulk Order

form

liquid

bp

238-242 °C

density

1.054 g/mL at 25 °C

functional group

amine

SMILES string

CC(C)(C)OC(=O)Nc1ccccc1CN

InChI

1S/C12H18N2O2/c1-12(2,3)16-11(15)14-10-7-5-4-6-9(10)8-13/h4-7H,8,13H2,1-3H3,(H,14,15)

InChI key

BFFRNSYPZZUZDR-UHFFFAOYSA-N

Application

Reactant for:
  • Two-step syntheses of fused quinoxaline-benzodiazepines and bis-benzodiazepines employing a double UDC (Ugi/Deprotect/Cyclize) strategy
  • Preparation of hydroquinazoline scaffolds via microwave-promoted Ugi reaction and cyclization
  • Preparation of pyrazinone inhibitors of mast cell tryptase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.1 °F - closed cup

Flash Point(C)

71.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service