Skip to Content
Merck
All Photos(1)

Key Documents

709204

Sigma-Aldrich

Hafnium boride

powder, −325 mesh, 99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HfB2
CAS Number:
Molecular Weight:
200.11
EC Number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: boron

reagent type: catalyst
core: hafnium

particle size

−325 mesh

mp

3100 °C

density

10.5 g/cm3 at 25 °C

SMILES string

[Hf]BB

InChI

1S/B2H3.Hf/c1-2;/h1H,2H2;/q+1;-1

InChI key

JLBVMPIAHRGRAQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service