Skip to Content
Merck
All Photos(2)

Key Documents

709123

Sigma-Aldrich

2-Methoxyethyl isothiocyanate

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$162.00

$162.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$162.00

About This Item

Empirical Formula (Hill Notation):
C4H7NOS
CAS Number:
Molecular Weight:
117.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$162.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.517

density

1.080 g/mL at 25 °C

functional group

amine
ether
isothiocyanate

SMILES string

COCCN=C=S

InChI

1S/C4H7NOS/c1-6-3-2-5-4-7/h2-3H2,1H3

InChI key

HTSAVXAFEVUJQE-UHFFFAOYSA-N

Related Categories

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.0 °F

Flash Point(C)

18.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service