Skip to Content
Merck
All Photos(2)

Key Documents

708127

Sigma-Aldrich

4-(Diphenylphosphino)styrene

97%

Synonym(s):

4-vinyltriphenylphosphine, DPPST, Diphenylphosphinostyrene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$42.90
5 G
$162.00
25 G
$397.00

$42.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$42.90
5 G
$162.00
25 G
$397.00

About This Item

Empirical Formula (Hill Notation):
C20H17P
CAS Number:
Molecular Weight:
288.32
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$42.90


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reagent type: polymerizable reagent

color

white to off-white

mp

75-80 °C

functional group

phosphine

SMILES string

C=Cc1ccc(cc1)P(c2ccccc2)c3ccccc3

InChI

1S/C20H17P/c1-2-17-13-15-20(16-14-17)21(18-9-5-3-6-10-18)19-11-7-4-8-12-19/h2-16H,1H2

InChI key

FRPLKHQCXVNBNO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service