Skip to Content
Merck
All Photos(1)

Key Documents

707600

Sigma-Aldrich

3-Benzyloxybenzoyl chloride

94%

Synonym(s):

3-Benzyloxybenzoic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11ClO2
CAS Number:
Molecular Weight:
246.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
solid
Assay:
94%

Assay

94%

form

solid

mp

40-43 °C

SMILES string

ClC(=O)c1cccc(OCc2ccccc2)c1

InChI

1S/C14H11ClO2/c15-14(16)12-7-4-8-13(9-12)17-10-11-5-2-1-3-6-11/h1-9H,10H2

InChI key

HPVUGOBQRQUWMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of dihydroquinolinones and tetrahydroquinazolinones as potent kinesin spindle protein inhibitors and potential antitumor agents
  • Preparation of carbamate derivatives as fatty acid amide hydrolase inhibitors

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service