Skip to Content
Merck
All Photos(4)

Key Documents

704954

Sigma-Aldrich

XPhos Pd G1

Synonym(s):

(2-Dicyclohexylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)[2-(2-aminoethyl)phenyl)]palladium(II) chloride, (XPhos) palladium(II) phenethylamine chloride, Chloro(2-dicyclohexylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)[2-(2-aminoethyl)phenyl)]palladium(II), XPhos Palladacycle, XPhos precatalyst

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$113.00
1 G
$324.00
25 G
$5,770.00

$113.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$113.00
1 G
$324.00
25 G
$5,770.00

About This Item

Empirical Formula (Hill Notation):
C41H59ClNPPd
CAS Number:
Molecular Weight:
738.76
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$113.00


Available to ship on22 April 2025Details


Request a Bulk Order

form

solid

Quality Level

feature

generation 1

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

205-210 °C

functional group

phosphine

SMILES string

NCCc1ccccc1[Pd]Cl.CC(C)c2cc(C(C)C)c(c(c2)C(C)C)-c3ccccc3P(C4CCCCC4)C5CCCCC5

InChI

1S/C33H49P.C8H10N.ClH.Pd/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28;9-7-6-8-4-2-1-3-5-8;;/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3;1-4H,6-7,9H2;1H;/q;;;+1/p-1

InChI key

HTAJCNQBAFZEJO-UHFFFAOYSA-M

General description

Material may contain up to 5% pentane

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Catalyst used for:
  • Amination / cyclization reactions
  • C-N bond-forming reactions via packed-bed microreactors
  • Boration of aryl chlorides
  • Arylation of oxazole
  • Cross-coupling reactions

Legal Information

Usage subject to US Patents 6307087 and 6395916.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service