Skip to Content
Merck
All Photos(1)

Key Documents

704830

Sigma-Aldrich

5-Bromo-2-(tributylstannyl)pyridine

97% (H-NMR)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30BrNSn
CAS Number:
Molecular Weight:
447.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97% (H-NMR)

form

liquid

refractive index

n20/D 1.535

bp

140-145 °C

density

1.317 g/mL at 25 °C

functional group

bromo

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccc(Br)cn1

InChI

1S/C5H3BrN.3C4H9.Sn/c6-5-2-1-3-7-4-5;3*1-3-4-2;/h1-2,4H;3*1,3-4H2,2H3;

InChI key

ZQJRPQMEYHJPJK-UHFFFAOYSA-N

Application

5-Bromo-2-(tributylstannyl)pyridine can be used in the synthesis of:
  • 1,10-Phenanthroline based ditopic ligand, which is used to prepare bistable metallic rotaxanes.[1]
  • Donor-acceptor type isomeric platinum(II) complexes exhibiting excellent optoelectronic properties for OLED applications.[2]
  • Pt(II) polymetallayne derived orange phosphorescent polymers applicable in phosphorescent OLEDs (PHOLEDs).[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancing the electroluminescence performances of novel platinum (ii) polymetallayne-based phosphorescent polymers through employing functionalized Ir III phosphorescent units and facilitating triplet energy transfer
Huang Z, et al.
Royal Society of Chemistry Advances, 5(16), 12100-12110 (2015)
Achieving NIR emission for donor-acceptor type platinum (II) complexes by adjusting coordination position with isomeric ligands
Zhang Y, et al.
Inorganic Chemistry, 57(22), 14208-14217 (2018)
Synthesis of a novel ditopic ligand incorporating directly bonded 1, 10-phenanthroline and 2, 2': 6', 2 ?-terpyridine units.
Gavi?a P and Tatay S.
Tetrahedron Letters, 47(20), 3471-3473 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service