Skip to Content
Merck
All Photos(1)

Documents

698776

Sigma-Aldrich

Chloro(tert-butyl)phenylphosphine

Synonym(s):

(1,1-Dimethylethyl)phenylphosphinous chloride, Chloro(phenyl)-tert-butylphosphine, tert-Butyl(phenyl)phosphinous chloride, tert-Butylchloro(phenyl)phosphine, tert-Butylphenylchlorophosphine, tert-butylphenylchlorophosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14ClP
CAS Number:
Molecular Weight:
200.64
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.555

bp

77-78 °C/0.4 mmHg

density

1.064 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)P(Cl)c1ccccc1

InChI

1S/C10H14ClP/c1-10(2,3)12(11)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

XOQDDCZARYSQNB-UHFFFAOYSA-N

Related Categories

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.0 °F

Flash Point(C)

32.8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Israel Cano et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(6), 1444-1450 (2016-11-23)
A specific secondary phosphine oxide (SPO) ligand (tert-butyl(phenyl)phosphine oxide) was employed to generate two iridium catalysts, an Ir-SPO complex and IrNPs (iridium nanoparticles) ligated with SPO ligands, which were compared mutually and with several supported iridium catalysts with the aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service