Skip to Content
Merck
All Photos(1)

Key Documents

698490

SAFC

1,2-Bis(dichlorophosphino)benzene

96%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$136.00

$136.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
250 MG
$136.00

About This Item

Empirical Formula (Hill Notation):
C6H4Cl4P2
CAS Number:
Molecular Weight:
279.86
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$136.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Assay

96%

form

liquid

refractive index

n20/D 1.662

density

1.545 g/mL at 25 °C

SMILES string

ClP(Cl)c1ccccc1P(Cl)Cl

InChI

1S/C6H4Cl4P2/c7-11(8)5-3-1-2-4-6(5)12(9)10/h1-4H

InChI key

IGYHSFVSIYJSML-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service