Skip to Content
Merck
All Photos(1)

Key Documents

698261

Sigma-Aldrich

Tris(4-methoxy-3,5-dimethylphenyl)phosphine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$52.70

$52.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$52.70

About This Item

Empirical Formula (Hill Notation):
C27H33O3P
CAS Number:
Molecular Weight:
436.52
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

$52.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

174-179 °C

functional group

phosphine

SMILES string

COc1c(C)cc(cc1C)P(c2cc(C)c(OC)c(C)c2)c3cc(C)c(OC)c(C)c3

InChI

1S/C27H33O3P/c1-16-10-22(11-17(2)25(16)28-7)31(23-12-18(3)26(29-8)19(4)13-23)24-14-20(5)27(30-9)21(6)15-24/h10-15H,1-9H3

InChI key

BDGUINGZRGNLPD-UHFFFAOYSA-N

Application

Catalyst for carbocyanation of alkynes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service