Skip to Content
Merck
All Photos(1)

Key Documents

698245

Sigma-Aldrich

Cyclohexyldichlorophosphine

95%

Synonym(s):

Cyclohexylphosphonous dichloride, Cyclohexylphosphorus dichloride, Dichlorocyclohexylphosphin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$89.10

$89.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$89.10

About This Item

Empirical Formula (Hill Notation):
C6H11Cl2P
CAS Number:
Molecular Weight:
185.03
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$89.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.528

density

1.204 g/mL at 25 °C

functional group

phosphine

SMILES string

ClP(Cl)C1CCCCC1

InChI

1S/C6H11Cl2P/c7-9(8)6-4-2-1-3-5-6/h6H,1-5H2

InChI key

MJEQIIGWDHUZJW-UHFFFAOYSA-N

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

153.0 °F

Flash Point(C)

67.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service