Skip to Content
Merck
All Photos(1)

Key Documents

698121

Sigma-Aldrich

4-Iodophenylboronic acid MIDA ester

97%

Synonym(s):

2-(4-Iodophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11BINO4
CAS Number:
Molecular Weight:
358.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

213-217 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccc(I)cc2

InChI

1S/C11H11BINO4/c1-14-6-10(15)17-12(18-11(16)7-14)8-2-4-9(13)5-3-8/h2-5H,6-7H2,1H3

InChI key

KIKNJEGAWBNFLW-UHFFFAOYSA-N

Application

4-Iodophenylboronic acid MIDA ester can be used:
  • As a starting material to prepare rod-like hydrogen-bonded organic frameworks through Stille-coupling using 2,5-bis(trimethyltin)heterocycle, Pd(PPh3)4, and CuI.
  • As a substrate in the C-H arylation reaction of pyroglutamic acid derivatives with aryl and heteroaryl iodides; catalyzed by the palladium catalyst.
  • As a substrate in the synthesis of trifluoroborates, which are further converted to protected boronic acids.
  • As a starting material in the synthesis of amino acid MIDA boronates as new building blocks by reacting with protected iodoalanine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Permanently porous hydrogen-bonded frameworks of rod-like thiophenes, selenophenes, and tellurophenes capped with MIDA boronates
Li P, et al.
Dalton Transactions, 45(24), 9754-9757 (2016)
A general method for interconversion of boronic acid protecting groups: Trifluoroborates as common intermediates
Churches QI, et al.
The Journal of Organic Chemistry, 80(11), 5428-5435 (2015)
Synthesis and properties of MIDA boronate containing aromatic amino acids: New peptide building blocks
Colgin N, et al.
Organic & Biomolecular Chemistry, 9(6), 1864-1870 (2011)
Stereospecific palladium-catalyzed C-H arylation of pyroglutamic acid derivatives at the C3 position enabled by 8-aminoquinoline as a directing group
Verho O, et al.
Organic Letters, 19(17), 4424-4427 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service