Skip to Content
Merck
All Photos(1)

Key Documents

697753

Sigma-Aldrich

(S,S)-DIPAMP

95%

Synonym(s):

(1S,2S)-(+)-Bis[(2-methoxyphenyl)phenylphosphino]ethane, (S,S)-1,2-Bis[(2-methoxyphenyl)(phenylphosphino)]ethane, (S,S)-1,2-Ethanediylbis[(2-methoxyphenyl)phenylphosphine]

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$147.00
500 MG
$462.00

$147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$147.00
500 MG
$462.00

About This Item

Linear Formula:
[CH3OC6H4P(C6H5)CH2]2
CAS Number:
Molecular Weight:
458.47
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

$147.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

optical activity

[α]22/D +82, c = 1 in chloroform

mp

101-105 °C (lit.)

functional group

phosphine

SMILES string

COc1ccccc1P(CCP(c2ccccc2)c3ccccc3OC)c4ccccc4

InChI

1S/C28H28O2P2/c1-29-25-17-9-11-19-27(25)31(23-13-5-3-6-14-23)21-22-32(24-15-7-4-8-16-24)28-20-12-10-18-26(28)30-2/h3-20H,21-22H2,1-2H3/t31-,32-/m0/s1

InChI key

QKZWXPLBVCKXNQ-ACHIHNKUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service