Skip to Content
Merck
All Photos(1)

Key Documents

695726

Sigma-Aldrich

(R)-3,3′-Di-9-anthracenyl-1,1′-bi-2-naphthol

97%

Synonym(s):

(R)-3,3′-Di-9-anthracenyl-[1,1′-binaphthalene]-2,2′-diol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$296.00

$296.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$296.00

About This Item

Empirical Formula (Hill Notation):
C48H30O2
CAS Number:
Molecular Weight:
638.75
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$296.00


Estimated to ship on15 April 2025


Request a Bulk Order

Quality Level

Assay

97%

form

powder

optical activity

[α]22/D +202°, c = 1 in chloroform

mp

125-130 °C

storage temp.

−20°C

SMILES string

Oc1c(cc2ccccc2c1-c3c(O)c(cc4ccccc34)-c5c6ccccc6cc7ccccc57)-c8c9ccccc9cc%10ccccc8%10

InChI

1S/C48H30O2/c49-47-41(43-35-19-7-1-13-29(35)25-30-14-2-8-20-36(30)43)27-33-17-5-11-23-39(33)45(47)46-40-24-12-6-18-34(40)28-42(48(46)50)44-37-21-9-3-15-31(37)26-32-16-4-10-22-38(32)44/h1-28,49-50H

InChI key

XJKZPSCYLJRHEN-UHFFFAOYSA-N

General description

Purity excludes solvent of crystallization. May contain up to 10% dichloromethane.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service