Skip to Content
Merck
All Photos(1)

Key Documents

695629

Sigma-Aldrich

5-Hexyl-2-thiopheneboronic acid pinacol ester

97%

Synonym(s):

2-(5-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$199.00
5 G
$661.00

$199.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 G
$199.00
5 G
$661.00

About This Item

Empirical Formula (Hill Notation):
C16H27BO2S
CAS Number:
Molecular Weight:
294.26
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$199.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5

bp

140-142 °C/0.5 mmHg

density

0.977 g/mL at 25 °C

SMILES string

CCCCCCc1ccc(s1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C16H27BO2S/c1-6-7-8-9-10-13-11-12-14(20-13)17-18-15(2,3)16(4,5)19-17/h11-12H,6-10H2,1-5H3

InChI key

FWZQTJFOKCBQGX-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service