Skip to Content
Merck
All Photos(1)

Key Documents

692336

Sigma-Aldrich

RuCl2[(R)−(DM−SEGPHOS®)][(R,(R))−(DPEN)]

Synonym(s):

Dichloro[(R)−5,5′−bis[di(3,5−xylyl)phosphino]−4,4′−bi−1,3−benzodioxole][(1R,2R)−1,2−diphenylethylenediamine]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H60Cl2N2O4P2Ru
CAS Number:
Molecular Weight:
1107.05
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

storage temp.

2-8°C

SMILES string

Cl[Ru]Cl.N[C@@H]([C@H](N)c1ccccc1)c2ccccc2.Cc3cc(C)cc(c3)P(c4cc(C)cc(C)c4)c5ccc6OCOc6c5-c7c8OCOc8ccc7P(c9cc(C)cc(C)c9)c%10cc(C)cc(C)c%10

InChI

1S/C46H44O4P2.C14H16N2.2ClH.Ru/c1-27-13-28(2)18-35(17-27)51(36-19-29(3)14-30(4)20-36)41-11-9-39-45(49-25-47-39)43(41)44-42(12-10-40-46(44)50-26-48-40)52(37-21-31(5)15-32(6)22-37)38-23-33(7)16-34(8)24-38;15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12;;;/h9-24H,25-26H2,1-8H3;1-10,13-14H,15-16H2;2*1H;/q;;;;+2/p-2/t;13-,14-;;;/m.1.../s1

InChI key

FOEKPXQPRAIJRW-ODQAEMFESA-L

Application

RuCl2[(R)-(DM-SEGPHOS®)][(R,(R))-(DPEN)] can be used as a catalyst in the enantioselective preparation of chiral amino alcohols and their derivatives by asymmetric hydrogenation of various amino ketones using mild hydrogen pressure.

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136 JP Application No. 19970359654
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric hydrogenation of amino ketones using chiral RuCl2 (diphophine)(1, 2-diamine) complexes
Ohkuma T, et a.
Journal of the American Chemical Society, 122(27), 6510-6511 (2000)

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service