Skip to Content
Merck
All Photos(1)

Key Documents

692220

Sigma-Aldrich

(R)-Ru(OAc)2(BINAP)

Synonym(s):

Diacetato[(R)-(+)-2,2′-bis(diphenylphosphino)-1,1′−binaphthyl]ruthenium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$36.70
500 MG
$154.00

$36.70


0 left in stock

Request a Bulk Order

Select a Size

Change View
100 MG
$36.70
500 MG
$154.00

About This Item

Empirical Formula (Hill Notation):
C48H38O4P2Ru
CAS Number:
Molecular Weight:
841.83
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$36.70


0 left in stock

Request a Bulk Order

form

powder

Quality Level

functional group

phosphine

storage temp.

2-8°C

SMILES string

CC(=O)O[Ru]OC(C)=O.c1ccc(cc1)P(c2ccccc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccccc7)c8ccccc8

InChI

1S/C44H32P2.2C2H4O2.Ru/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;2*1-2(3)4;/h1-32H;2*1H3,(H,3,4);/q;;;+2/p-2

InChI key

NMLZYEWNUCRSRJ-UHFFFAOYSA-L

Application

(R)-Ru(OAc)2(BINAP) may be used as a catalyst for the asymmetric hydrogenation of 1,1′-disubstituted olefins, such as 1-methyleneindan and 1-methylenetetralins.[1] It can also catalyze the hydrogenation of 2-(trifluoromethyl)acrylic acid to form optically active 2-(trifluoromethyl)propionic acid.[2]
Catalyst for:
  • Asymmetric hydrogenation reactions

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BINAP-Ru (II) and BINAP-Rh (I)-catalyzed asymmetric hydrogenation of olefins without heteroatom-functionalities
Ohta T, et al.
Journal of Organometallic Chemistry, 502(1-2), 169-176 (1995)
Preparation of optically active 2-(trifluoromethyl) alkan-1-ols by catalytic asymmetric hydrogenation.
Iseki K, et al.
Journal of Fluorine Chemistry, 69(1), 5-6 (1994)

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service