Skip to Content
Merck
All Photos(1)

Key Documents

69180

Sigma-Aldrich

2-Methylstyrene

≥95.0% (GC)

Synonym(s):

2-Vinyltoluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$274.00

$274.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$274.00

About This Item

Empirical Formula (Hill Notation):
C9H10
CAS Number:
Molecular Weight:
118.18
Beilstein:
1901764
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$274.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.543

bp

169-171 °C

density

0.914 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cc1ccccc1C=C

InChI

1S/C9H10/c1-3-9-7-5-4-6-8(9)2/h3-7H,1H2,2H3

InChI key

NVZWEEGUWXZOKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kühler
Xenobiotica; the fate of foreign compounds in biological systems, 14(5), 417-428 (1984-05-01)
N-Acetyl-L-cysteine was reacted with 2-(2-, 3-, or 4-methylphenyl)-oxiranes to give mixtures of the two possible regio isomers N-acetyl-S-[1-(2-, 3-, or 4-methylphenyl)-2-hydroxyethyl]-L-cysteine and N-acetyl-S-[2-(2-, 3-, or 4-methylphenyl)-2-hydroxyethyl]-L-cysteine, respectively. These were isolated in pure form by h.p.l.c.. The diastereomers were characterized by
Yasuhito Suzuki et al.
Forensic science international, 278, 228-239 (2017-08-02)
Various autopsy findings are used for the diagnosis of burning death, including inhalation injury, soot in the respiratory tract, vital reaction, blood carboxyhemoglobin (COHb), and blood cyanide. However, not all findings are necessarily complete, and autopsy diagnosis can be difficult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service