Skip to Content
Merck
All Photos(3)

Key Documents

691550

Sigma-Aldrich

N-Boc-tyramine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$201.00
25 G
$602.00

$201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$201.00
25 G
$602.00

About This Item

Empirical Formula (Hill Notation):
C13H19NO3
CAS Number:
Molecular Weight:
237.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$201.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

71-75 °C

functional group

amine

SMILES string

CC(C)(C)OC(=O)NCCc1ccc(O)cc1

InChI

1S/C13H19NO3/c1-13(2,3)17-12(16)14-9-8-10-4-6-11(15)7-5-10/h4-7,15H,8-9H2,1-3H3,(H,14,16)

InChI key

ILNOTKMMDBWGOK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faezeh Sharifi et al.
Journal of colloid and interface science, 585, 126-137 (2020-12-07)
Simple zwitterions used as auxiliary agents might have the potential to change the zeta potential of oil-in-water (o/w) nanoemulsions on the mucosa. The zwitterion phosphorylated tyramine (p-Tyr) was synthesized by phosphorylation of Boc-tyramine (Boc-Tyr) using phosphoryl chloride (POCl3). It was
Chien-Wei Feng et al.
Marine drugs, 17(6) (2019-05-31)
Pharmaceutical agents for halting the progression of Parkinson's disease (PD) are lacking. The current available medications only relieve clinical symptoms and may cause severe side effects. Therefore, there is an urgent need for novel drug candidates for PD. In this
Chutima Talabnin et al.
Oncology letters, 20(3), 2483-2492 (2020-08-13)
Piperlongumine (PL) produces reactive oxygen species (ROS) and induces G2/M-phase arrest in cholangiocarcinoma (CCA) cells via the JNK/ERK pathway. A differential response to PL was observed among all CCA cell lines However, the underlying mechanisms have remained to be fully
Leonie Zeitler et al.
eLife, 10 (2021-03-02)
Interleukin-4-induced-1 (IL4i1) is an amino acid oxidase secreted from immune cells. Recent observations have suggested that IL4i1 is pro-tumorigenic via unknown mechanisms. As IL4i1 has homologs in snake venoms (L-amino acid oxidases [LAAO]), we used comparative approaches to gain insight

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service