Skip to Content
Merck
All Photos(1)

Key Documents

691097

Sigma-Aldrich

1,5-Cyclooctadiene{[dibenzyl((4S,5S)-5-methyl-2-phenyl-4,5-dihydro-4-oxazolyl)methyl]diphenylphosphinite κN:κP}iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)borate

97%

Synonym(s):

ThrePHOX, Ubaphox, [((4S,5S)-Ph2-Ubaphox)Ir(COD)]BARF

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$148.00
5 G
$4,120.00

$148.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$148.00
5 G
$4,120.00

About This Item

Empirical Formula (Hill Notation):
C77H58BF24IrNO2P
CAS Number:
Molecular Weight:
1719.25
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$148.00


Estimated to ship on15 April 2025


Request a Bulk Order

Quality Level

Assay

97%

form

lumps

optical purity

ee: ≥99%

storage temp.

2-8°C

SMILES string

[Ir+].[CH]1[CH]CC[CH][CH]CC1.C[C@@H]2OC(=N[C@@H]2C(Cc3ccccc3)(Cc4ccccc4)OP(c5ccccc5)c6ccccc6)c7ccccc7.FC(F)(F)c8cc(cc(c8)C(F)(F)F)[B-](c9cc(cc(c9)C(F)(F)F)C(F)(F)F)(c%10cc(cc(c%10)C(F)(F)F)C(F)(F)F)c%11cc(cc(c%11)C(F)(F)F)C(F)(F)F

InChI

1S/C37H34NO2P.C32H12BF24.C8H12.Ir/c1-29-35(38-36(39-29)32-21-11-4-12-22-32)37(27-30-17-7-2-8-18-30,28-31-19-9-3-10-20-31)40-41(33-23-13-5-14-24-33)34-25-15-6-16-26-34;34-25(35,36)13-1-14(26(37,38)39)6-21(5-13)33(22-7-15(27(40,41)42)2-16(8-22)28(43,44)45,23-9-17(29(46,47)48)3-18(10-23)30(49,50)51)24-11-19(31(52,53)54)4-20(12-24)32(55,56)57;1-2-4-6-8-7-5-3-1;/h2-26,29,35H,27-28H2,1H3;1-12H;1-2,7-8H,3-6H2;/q;-1;;+1/b;;2-1-,8-7-;/t29-,35-;;;/m0.../s1

InChI key

RWJSSAILCUYYLD-RDSUWHNOSA-N

General description

sold in collaboration with Solvias AG

Application

Catalyst for enantioselective hydrogenation of alkenes[1][2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New Class of Modular Phosphinite-Oxazoline Ligands: Ir-Catalyzed Enantioselective Hydrogenation of Alkenes We thank Dr. Martin Studer and Dr. Benoît Pugin (Solvias AG, Basel) for fruitful discussions, and Prof. Kevin Burgess (Texas A&M University) for preprints of related unpublished work. Financial support by the Swiss National Science Foundation is gratefully acknowledged.
Jörg Blankenstein et al.
Angewandte Chemie (International ed. in English), 40(23), 4445-4447 (2002-10-31)
Threonine-derived phosphinite-oxazoline ligands for the Ir-catalyzed enantioselective hydrogenation.
Menges, F. and Pfaltz, A.
Advanced Synthesis & Catalysis, 344, 40-44 (2002)

Related Content

Andreas Pfaltz's chiral ligand designs, like semicorrins, pioneered bisoxazolines for widespread catalytic asymmetric synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service