Skip to Content
Merck
All Photos(1)

Key Documents

690821

Sigma-Aldrich

1,3-Dimethoxyimidazolium hexafluorophosphate

98%, complies for IR spectroscopy

Synonym(s):

(OMe)2Im-PF6

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$164.00

$164.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$164.00

About This Item

Empirical Formula (Hill Notation):
C5H9F6N2O2P
CAS Number:
Molecular Weight:
274.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$164.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (13C-NMR)
≥98% (31P-NMR)
98%

form

solid

mp

84 °C

suitability

complies for IR spectroscopy

SMILES string

F[P-](F)(F)(F)(F)F.COn1cc[n+](OC)c1

InChI

1S/C5H9N2O2.F6P/c1-8-6-3-4-7(5-6)9-2;1-7(2,3,4,5)6/h3-5H,1-2H3;/q+1;-1

InChI key

KBRFXVGODOTNGL-UHFFFAOYSA-N

Application

1,3-Dimethoxyimidazolium hexafluorophosphate can be used:
  • As an ionic liquid and N-heterocyclic carbene (NHC) precatalyst.[1]
  • To prepare bis(1,3-dimethoxyimidazolin-2-ylidene)silver(I) hexafluoridophosphate complex, which acts as an N-heterocyclic carbene.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N, N′-Di (alkyloxy) imidazolium salts: New patent-free ionic liquids and NHC precatalysts
Laus G, et al.
Zeitschrift fur Naturforschung B, 62(3), 295-308 (2007)
Crystal structure of bis (1, 3-dimethoxyimidazolin-2-ylidene) silver (I) hexafluoridophosphate, N-heterocyclic carbene (NHC) complex
Rietzler B, et al.
Acta Crystallographica Section E: Crystallographic Communications, 71(12), m251-m252 (2015)
N,N-Di(alkoxy)imidazolium salts: new patent-free ionic liquids and NHC-precatalysts
Laus, G., et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 62, 295-308 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service