Skip to Content
Merck
All Photos(1)

Key Documents

689424

Sigma-Aldrich

Dehydroacetic acid sodium salt monohydrate

Arxada quality, 98.0-101.0 % (w/w) (T)

Synonym(s):

3-Acetyl-4-hydroxy-6-methyl-2-pyrone sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NaO4 · H2O
CAS Number:
Molecular Weight:
208.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

quality

Arxada quality

manufacturer/tradename

Arxada AG

concentration

98.0-101.0 % (w/w) (T)

impurities

8.2-9.5 % (w/w) water (Karl Fischer)

SMILES string

O.[Na+].CC(=O)C1=C([O-])C=C(C)OC1=O

InChI

1S/C8H8O4.Na.H2O/c1-4-3-6(10)7(5(2)9)8(11)12-4;;/h3,10H,1-2H3;;1H2/q;+1;/p-1

InChI key

XBTXRILZQWUAPE-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huizhen Zhang et al.
Wei sheng yan jiu = Journal of hygiene research, 36(6), 748-750 (2008-02-29)
To estabish a high performance liquid chromatographic (HPLC) method for the determination of dehydroaetic acid in food determinated. Separation was carried out on a column of C18 (51 microm, 250 x 4.60mm) at the temperature of 25 degrees C with
Allergic contact dermatitis caused by sodium dehydroacetate, not hyaluronic acid, in Ialuset® cream.
Brigitte Milpied et al.
Contact dermatitis, 65(6), 359-361 (2011-11-15)
Luiz C Dias et al.
Journal of agricultural and food chemistry, 57(4), 1399-1405 (2009-01-28)
A short and efficient approach to a range of new chiral and achiral functionalized (E)-enaminopyran-2,4-diones starting with commercially available dehydroacetic acid is described. The phytotoxic properties of these (E)-enaminopyran-2,4-diones were evaluated by their ability to interfere with the growth of
C S Mantzoros et al.
American journal of hypertension, 8(6), 606-614 (1995-06-01)
To test the hypothesis that androgens may play a role in the pathogenesis of atherosclerosis and hypertension, we investigated the association of delta 5 dehydroepiandrosterone sulfate [DHEAS]) and delta 4 (free testosterone [FT]) androgens with the resting, seated blood pressure
Yuka Sakaguchi et al.
Journal of applied toxicology : JAT, 28(4), 524-529 (2007-10-18)
Sodium dehydroacetate (DHA-S) is used as a food additive, preservative and antimicrobial agent. Repeated oral administration of DHA-S in rats induced severe hemorrhage in multiple organs and prolongation of blood coagulation factors. To determine the mechanism of hemorrhage, the protective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service