Skip to Content
Merck
All Photos(1)

Key Documents

688053

Sigma-Aldrich

Tris(4-carbazoyl-9-ylphenyl)amine

97%

Synonym(s):

TCTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H36N4
CAS Number:
Molecular Weight:
740.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

179-183 °C

Orbital energy

HOMO 5.7 eV 
LUMO 2.4 eV 

OLED Device Performance

ITO/NPD/TCTA/BCPO:Ir(piq)3 (7-8%)/BCP/Alq3/LiF/Al

  • Color: red
  • Max. Luminance: 24529 Cd/m2
  • Max. EQE: 17 %
  • Turn-On Voltage: 2.7 V

ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: green
  • Max. Luminance: 207839 Cd/m2
  • Max. EQE: 21.6 %
  • Turn-On Voltage: 2.1 V

ITO/PEDOT:PSS/NPD/TCTA/m-CBTZ:FIrpic (10 wt%)/DPPS/TAZ/LiF/Al
  • Color: blue
  • Max. Luminance: 29500 Cd/m2
  • Max. EQE: 8.8 %
  • Turn-On Voltage: 3.5 V

ITO/PEDOT:PSS/NPD/TCTA/m-CBTZ:Ir(Bt)2(acac) (10%)/TPBI/LiF/Al
  • Color: yellow
  • Max. Luminance: 106000 Cd/m2
  • Max. EQE: 17.5 %
  • Turn-On Voltage: 3 V

SMILES string

c1ccc2c(c1)n(-c3ccc(cc3)N(c4ccc(cc4)-n5c6ccccc6c7ccccc57)c8ccc(cc8)-n9c%10ccccc%10c%11ccccc9%11)c%12ccccc2%12

InChI

1S/C54H36N4/c1-7-19-49-43(13-1)44-14-2-8-20-50(44)56(49)40-31-25-37(26-32-40)55(38-27-33-41(34-28-38)57-51-21-9-3-15-45(51)46-16-4-10-22-52(46)57)39-29-35-42(36-30-39)58-53-23-11-5-17-47(53)48-18-6-12-24-54(48)58/h1-36H

InChI key

AWXGSYPUMWKTBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OLED hole transporter material.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Inyoung Song et al.
Korean journal of radiology, 19(5), 905-915 (2018-09-04)
To compare the diagnostic performance of electrocardiogram (ECG)-gated thoracic computed tomography angiography (TCTA) without heart rate (HR) control in ischemic stroke patients with coronary CTA (CCTA) in non-stroke patients for detection of significant coronary artery stenosis. From September 2009 through
Brigita Vigante et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(13), 3325-3336 (2018-12-12)
With the aim of developing all-organic bipolar semiconductors with high charge mobility and efficient E-type fluorescence (so-called TADF) as environmentally friendly light-emitting materials for optoelectronic applications, four noble metals-free dyes with linear and V-shapes were designed using accepting pyridine-3,5-dicarbonitrile and
Xue Bai et al.
Electrophoresis, 40(12-13), 1662-1676 (2019-04-24)
We have developed a novel STR 25-plex florescence multiplex-STR kit (DNATyper25) to genotype 23 autosomal and two sex-linked loci for forensic applications and paternity analysis. Of the 23 autosomal loci, 20 are non-CODIS. The sex-linked markers include a Y-STR locus
Jamie L Almeida et al.
Cytotechnology, 66(1), 133-147 (2013-02-23)
The scientific community has responded to the misidentification of human cell lines with validated methods to authenticate these cells; however, few assays are available for nonhuman cell line identification. We have developed a multiplex polymerase chain reaction assay that targets
Markus Fröbel et al.
Scientific reports, 8(1), 9684-9684 (2018-06-28)
In recent years, the organic light-emitting diode (OLED) technology has been a rapidly evolving field of research, successfully making the transition to commercial applications such as mobile phones and other small portable devices. OLEDs provide efficient generation of light, excellent

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service