Skip to Content
Merck
All Photos(2)

Key Documents

687847

Sigma-Aldrich

Quinoxaline N-oxide

97%

Synonym(s):

1-Oxide quinoxaline

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$316.00

$316.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$316.00

About This Item

Empirical Formula (Hill Notation):
C8H6N2O
CAS Number:
Molecular Weight:
146.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$316.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

121-125 °C

SMILES string

[O-][n+]1ccnc2ccccc12

InChI

1S/C8H6N2O/c11-10-6-5-9-7-3-1-2-4-8(7)10/h1-6H

InChI key

OARGFWQSVACNCO-UHFFFAOYSA-N

Application

Coupling partner in Pd-catalyzed direct arylation reactions.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Campeau, L. C. et al.
Aldrichimica Acta, 40, 35-35 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service