Skip to Content
Merck
All Photos(1)

Key Documents

687332

Sigma-Aldrich

N-Boc-trans-4-fluoro-L-proline

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$184.00

$184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$184.00

About This Item

Empirical Formula (Hill Notation):
C10H16FNO4
CAS Number:
Molecular Weight:
233.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$184.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

optical activity

[α]22/D -65.0±5°, c = 1 in chloroform

mp

115-119 °C

functional group

carboxylic acid
fluoro

SMILES string

CC(C)(C)OC(=O)N1C[C@H](F)C[C@H]1C(O)=O

InChI

1S/C10H16FNO4/c1-10(2,3)16-9(15)12-5-6(11)4-7(12)8(13)14/h6-7H,4-5H2,1-3H3,(H,13,14)/t6-,7+/m1/s1

InChI key

YGWZXQOYEBWUTH-RQJHMYQMSA-N

Application

N-Boc-trans-4-fluoro-L-proline can be used in the preparation of benzophenone based ligand, which is further used to synthesize Ni(II) Schiff base complex.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Ni (II) Schiff base complexation and structural analysis of fluorinated analogs of the ligand (S)-2-[N-(N′-benzylprolyl) amino] benzophenone (BPB)
Tatum NJ, et al.
Journal of Fluorine Chemistry, 173, 77-83 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service