Skip to Content
Merck
All Photos(1)

Documents

686603

Sigma-Aldrich

Hyperbranched bis-MPA polyester-16-hydroxyl, generation 2

≥97%

Synonym(s):

ALH-16-OH, Hyperbranched Polymer, Hyperpolymer, PFH-16-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C75H128O45
CAS Number:
Molecular Weight:
1749.79
UNSPSC Code:
12162002
NACRES:
NA.23

description

hydroxyl surface groups

Assay

≥97%

form

powder (lyophilized)

mol wt

generation 2

no. Surface Groups

16 (avg.)

storage temp.

2-8°C

SMILES string

CC(CO)(CO)C(=O)OCC(C)(COC(=O)C(C)(CO)CO)C(=O)OCCOCCOCC(COCCOC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)(COCCOC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COCCOC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO

General description

Soluble in polar solvents such as DMSO, and methanol. Emulsifies in water (colloidal system).

Other Notes

bis-MPA = 2,2-bis(hydroxymethyl)propionic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dendrons in nanomaterials - Find features and benefits of polyester dendrons and hyperbranched polymer products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service