Skip to Content
Merck
All Photos(1)

Key Documents

68571

Sigma-Aldrich

Aminomethyl ChemMatrix® resin

0.50-0.70 mmol/g loading (amine)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

product line

ChemMatrix®

form

crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

particle size

100-200 mesh

capacity

0.50-0.70 mmol/g loading (amine)

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

Aminomethyl ChemMatrix® resin is a polyethyleneglycol (PEG)-based resin, composed of highly cross-linked primary ether bonds, therefore the resin is chemically and mechanically stable in a broad range of solvents. It has good loading properties when compared to other PEG resins due to its availability in free-flowing powder form. This resin is also more effective than polystyrene resins in the synthesis of hydrophobic or highly structured polypeptides.[1][2]

Application

Aminomethyl ChemMatrix® resin can be used as efficient support in solid-phase peptide synthesis (SPPS).[2][3] It is also used as a starting material to synthesize dihydrolipoyl-supported polymer, which is used as an efficient and recyclable reducing agent for the reduction of disulfide compounds.[4]

Legal Information

ChemMatrix is a registered trademark of Matrix Innovation, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convenient supported recyclable material based on dihydrolipoyl-residue for the reduction of disulfide derivatives
Bienvenu C, et al.
Tetrahedron Letters, 51(25), 3309-3311 (2010)
Polyethyleneglycol-based resins as solid supports for the synthesis of difficult or long peptides
Beatriz G, et al.
International Journal of Peptide Research and Therapeutics, 13(1-2), 265-270 (2007)
Solid-phase peptide synthesis using acetonitrile as a solvent in combination with PEG-based resins
Gerardo A Acosta, et al.
Journal of Peptide Science, 15(10), 629-633 (2009)
ChemMatrix for complex peptides and combinatorial chemistry.
Garcia-Ramos Y, et al.
Journal of Peptide Science, 16(12), 675-678 (2010)

Articles

ChemMatrix® is a proprietary, 100% PEG (polyethylene glycol) based resin from PCAS BioMatrix.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service