Skip to Content
Merck
All Photos(1)

Key Documents

685569

Sigma-Aldrich

(11bS)-N,N-dimethyl-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine

Synonym(s):

MonoPhos Ligand

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$219.00

$219.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$219.00

About This Item

Empirical Formula (Hill Notation):
C22H26NO2P
CAS Number:
Molecular Weight:
367.42
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$219.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

optical activity

[α]22/D 282.0°, c = 1 in chloroform

mp

86-94 °C

functional group

amine

SMILES string

CN(C)P1Oc2ccc3CCCCc3c2-c4c(O1)ccc5CCCCc45

InChI

1S/C22H26NO2P/c1-23(2)26-24-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)25-26/h11-14H,3-10H2,1-2H3

InChI key

OIZQADYWBXZQKE-UHFFFAOYSA-N

Application

(11bS)-N,N-dimethyl-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine is a monodentate phosphoramidite (MonoPhos) chiral ligand that can be used:
  • In the Cu-catalyzed synthesis of chiral 3-(arylbut-1-yn-1-yl)silane derivatives by reacting N-tosylhydrazones with silyl-substituted alkynes via asymmetric insertion.[1]
  • In the Rh-catalyzed asymmetric hydrogenation of R-isopropylcinnamic acid derivative, a key intermediate of renin inhibitor aliskiren.[2]
  • In the Rh-catalyzed conjugate addition of arylboronic acids to lactones, enones, and nitroalkenes.[3]

Legal Information

Sold under license from DSM for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mixed-ligand approach enables the asymmetric hydrogenation of an α-isopropylcinnamic acid en route to the renin inhibitor aliskiren
Boogers JAF, et al.
Organic Process Research & Development, 11(3), 585-591 (2007)
Asymmetric Copper-Catalyzed C (sp)-H Bond Insertion of Carbenoids Derived from N-Tosylhydrazones
Osako T, et al.
Synlett, 29(17), 2251-2256 (2018)
Rhodium-catalyzed asymmetric conjugate additions of boronic acids using monodentate phosphoramidite ligands
Boiteau J, et al.
Organic Letters, 5(5), 681-684 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service