Skip to Content
Merck
All Photos(3)

Documents

684821

Sigma-Aldrich

1,2-Bis(phenylsulfinyl)ethane palladium(II) acetate

greener alternative

Synonym(s):

White catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20O6PdS2
CAS Number:
Molecular Weight:
502.90
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: C-H Activation

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

SMILES string

CC(=O)O[Pd]OC(C)=O.O=S(CCS(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H14O2S2.2C2H4O2.Pd/c15-17(13-7-3-1-4-8-13)11-12-18(16)14-9-5-2-6-10-14;2*1-2(3)4;/h1-10H,11-12H2;2*1H3,(H,3,4);/q;;;+2/p-2

InChI key

SNNYSJNYZJXIFE-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Application

Catalyst for allylic C-H oxidation, useful for preparation of branched allylic esters, macrocycles, and amino alcohol derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

syn-1,2-Amino alcohols via diastereoselective allylic C-H amination.
Kenneth J Fraunhoffer et al.
Journal of the American Chemical Society, 129(23), 7274-7276 (2007-05-23)
Kenneth J Fraunhoffer et al.
Journal of the American Chemical Society, 128(28), 9032-9033 (2006-07-13)
A novel Pd/sulfoxide-catalyzed macrolactonization reaction of linear omega-alkenoic acids is reported that proceeds via serial ligand-catalyzed allylic C-H oxidation. The scope of this macrolactonization appears to be very broad. Aryl, alkyl, and (Z)-alpha,beta-unsaturated acids are all competent nucleophiles for this

Articles

The "White catalyst" by Professor M. Christina White enables allylic carbon functionalization, demonstrating exceptional catalytic activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service