Skip to Content
Merck
All Photos(1)

Key Documents

684678

Sigma-Aldrich

2,2-Difluorocyclopropanecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$151.00

$151.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$151.00

About This Item

Empirical Formula (Hill Notation):
C4H4F2O2
CAS Number:
Molecular Weight:
122.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$151.00


Estimated to ship on12 April 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

bp

70 °C/7 mmHg

mp

60-65 °C

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)C1CC1(F)F

InChI

1S/C4H4F2O2/c5-4(6)1-2(4)3(7)8/h2H,1H2,(H,7,8)

InChI key

KMLMOVWSQPHQME-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service