Skip to Content
Merck
All Photos(1)

Key Documents

684406

Sigma-Aldrich

1,1′-Bis[(2S,5S)-2,5-diisopropylphospholano]ferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48FeP2
CAS Number:
Molecular Weight:
526.49
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

form

solid

mp

69.1-73.0 °C

SMILES string

[Fe].CC(C)[C@@H]1CC[C@@H](C(C)C)P1[C]2[CH][CH][CH][CH]2.CC(C)[C@@H]3CC[C@@H](C(C)C)P3[C]4[CH][CH][CH][CH]4

InChI

1S/2C15H24P.Fe/c2*1-11(2)14-9-10-15(12(3)4)16(14)13-7-5-6-8-13;/h2*5-8,11-12,14-15H,9-10H2,1-4H3;/t2*14-,15-;/m00./s1

InChI key

CIIWEDVVKXIYFL-XRLMYZCVSA-N

Application

DuPhos and BPE Ligands: Highly Efficient Privileged Ligands

  • Chiral catalystic ligand used for enantioselective rhodium-catalyzed hydrogenation reactions

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service