Skip to Content
Merck
All Photos(1)

Key Documents

683981

Sigma-Aldrich

(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H, 6H-indeno[2,1-b]-1,2,4-triazolo[4,3-d]-1,4-oxazinium chloride monohydrate

97%

Synonym(s):

Bode Catalyst 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClN3O · H2O
Molecular Weight:
385.89
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

optical activity

[α]20/D -156.0°, c = 1 in chloroform

mp

212-216 °C

functional group

ether

SMILES string

[Cl-].Cc1cc(C)c(c(C)c1)-[n+]2c[n@H]3[C@H]4[C@H](Cc5ccccc45)OCc3n2

InChI

1S/C21H22N3O.ClH/c1-13-8-14(2)20(15(3)9-13)24-12-23-19(22-24)11-25-18-10-16-6-4-5-7-17(16)21(18)23;/h4-9,12,18,21H,10-11H2,1-3H3;1H/q+1;/p-1/t18-,21+;/m0./s1

InChI key

GUECWMLEUCWYOS-OZYANKIXSA-M

Application

(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H, 6H-indeno[2,1-b]-1,2,4-triazolo[4,3-d]-1,4-oxazinium chloride monohydrate can be used as an N-heterocyclic carbene (NHC) organocatalyst:
  • In the synthesis of tricyclic β-lactones by reacting 2-bromoenals with 1,3-diketones.[1]
  • To functionalize the C2-position of 3-aminobenzofurans using 2-bromoenals as coupling partners.[2]
  • In the synthesis of dihydroquinoxalines from substituted bisimines.[3]
  • In the preparation of spiro-glutarimide derivatives by reacting cyclic β-ketoamides with α,β-unsaturated aldehydes.[4]

Legal Information

Sold in collaboration with BioBlocks, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Synthesis of Tricyclic β-Lactones by NHC-Catalyzed Desymmetrization of Cyclic 1, 3-Diketones
Shee S, et al.
Organic Letters, 22(14), 5407?5411-5407?5411 (2020)
Catalytic, Enantioselective C2-Functionalization of 3-Aminobenzofurans Using N-Heterocyclic Carbenes
Barik S, et al.
Organic Letters, 22(10), 3865?3869-3865?3869 (2020)
N-Heterocyclic Carbene-Catalyzed Umpolung of Imines for the Enantioselective Synthesis of Dihydroquinoxalines
Das TK, et al.
ACS Catalysis, 9(5), 4065-4071 (2019)
N-heterocyclic carbene-catalyzed enantioselective synthesis of spiro-glutarimides via α, β-unsaturated acylazoliums
Mondal S, et al.
Organic Letters, 20(15), 4499-4503 (2018)

Related Content

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service