Skip to Content
Merck
All Photos(1)

Key Documents

682616

Sigma-Aldrich

Nanoclay, surface modified

contains 25-30 wt. % octadecylamine

Synonym(s):

Nanomer® I.30E, Nanomer® clay, Montmorillonite clay

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100

form

powder

contains

25-30 wt. % octadecylamine

matrix

Montmorillonite clay base material

particle size

≤20 μm

Looking for similar products? Visit Product Comparison Guide

Application

For representative application data and nanocomposite processing methods see datasheets T11 and T13.
Incorporated in amine-cured epoxy polyurethanes
  • Speed up cure catalyst
  • Improve modulus
  • Increase Tg
  • Enhance chemical resistance

Legal Information

Nanomer® clays are products of Nanocor Corporation
Product of Nanocor®, Inc.
Nanomer is a registered trademark of Amcol International Corporation

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service