Skip to Content
Merck
All Photos(3)

Key Documents

682373

Sigma-Aldrich

Hoveyda-Grubbs Catalyst® M721

Umicore

Synonym(s):

Hoveyda-Grubbs Catalyst® M72 SI(o-Tol) (C571), Dichloro[1,3-bis(2-methylphenyl)-2-imidazolidinylidene](2-isopropoxyphenylmethylene)ruthenium(II), Stewart-Grubbs catalyst

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$154.00
500 MG
$730.00
2 G
$2,060.00

$154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$154.00
500 MG
$730.00
2 G
$2,060.00

About This Item

Empirical Formula (Hill Notation):
C27H30Cl2N2ORu
CAS Number:
Molecular Weight:
570.52
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$154.00


Check Cart for Availability

Request a Bulk Order

Quality Level

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

mp

199-208 °C

storage temp.

2-8°C

SMILES string

CC(C)Oc1ccccc1C=[Ru](Cl)(Cl)=C2N(CCN2c3ccccc3C)c4ccccc4C

InChI

1S/C17H18N2.C10H12O.2ClH.Ru/c1-14-7-3-5-9-16(14)18-11-12-19(13-18)17-10-6-4-8-15(17)2;1-8(2)11-10-7-5-4-6-9(10)3;;;/h3-10H,11-12H2,1-2H3;3-8H,1-2H3;2*1H;/q;;;;+2/p-2

InChI key

ZDJCARGGZRTYFH-UHFFFAOYSA-L

Application

Less sterically hindered analog of Hoveyda-Grubbs Catalyst® 2nd Generation. Excels at ring-closing metathesis reactions of sterically encumbered olefins. Also useful for the cross metathesis of olefins bearing large allylic substituents.

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David E White et al.
Journal of the American Chemical Society, 130(3), 810-811 (2008-01-01)
Described in this report is the first total synthesis of elatol, a halogenated sesquiterpene in the chamigrene natural product family. The key disconnections in our synthetic approach include an enantioselective decarboxylative allylation to form the all-carbon quaternary stereocenter and a
Ian C Stewart et al.
Organic letters, 9(8), 1589-1592 (2007-03-24)
[reaction: see text] A series of ruthenium-based metathesis catalysts with N-heterocyclic carbene (NHC) ligands have been prepared in which the N-aryl groups have been changed from mesityl to mono-ortho-substituted phenyl (e.g., tolyl). These new catalysts offer an exceptional increase in
Schrodi, Y.; Pederson, R.L.
Aldrichimica Acta, 40, 45-45 (2007)

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service