Skip to Content
Merck
All Photos(2)

Key Documents

682071

Sigma-Aldrich

2-(Hydroxymethyl)phenylboronic acid cyclic monoester

97%

Synonym(s):

1,3-Dihydro-1-hydroxy-2,1-benzoxaborole, 2-(Hydroxymethyl)phenylboronic acid hemiester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$89.80
5 G
$379.00

$89.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$89.80
5 G
$379.00

About This Item

Empirical Formula (Hill Notation):
C7H7BO2
CAS Number:
Molecular Weight:
133.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$89.80


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

95-100 °C

SMILES string

OB1OCc2ccccc12

InChI

1S/C7H7BO2/c9-8-7-4-2-1-3-6(7)5-10-8/h1-4,9H,5H2

InChI key

XOQABDOICLHPIS-UHFFFAOYSA-N

Application

Oxaboroles and benzoxaboroles are useful substrates to prepare allylic and benzylic alcohols via Suzuki coupling.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 1148-1148 (2006)
Eric C Hansen et al.
Journal of the American Chemical Society, 128(25), 8142-8143 (2006-06-22)
A novel mode of regiochemical control over the allylic [1,3]-transposition of silyloxy groups catalyzed by Re2O7 has been developed. This strategy relies on a cis-oriented vinyl boronate, generated from the Alder-ene reaction of homoallylic silyl ethers and alkynyl boronates, to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service