Skip to Content
Merck
All Photos(1)

Key Documents

681903

Sigma-Aldrich

Indoline-7-carboxaldehyde

97%

Synonym(s):

2,3-Dihydro-1H-indole-7-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

46-50 °C

SMILES string

[H]C(=O)c1cccc2CCNc12

InChI

1S/C9H9NO/c11-6-8-3-1-2-7-4-5-10-9(7)8/h1-3,6,10H,4-5H2

InChI key

SJROBXXAXJAJOG-UHFFFAOYSA-N

Application

Useful starter in indoline chemistry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 9, 124-124 (1998)
Organic Syntheses, 73, 85-85 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service