Skip to Content
Merck
All Photos(2)

Key Documents

680214

Sigma-Aldrich

N-Boc-4-piperidineacetaldehyde

97%

Synonym(s):

4-(2-Oxoethyl)piperidine-1-carboxylic acid, tert-butyl ester, tert-Butyl 4-(formylmethyl)piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO3
CAS Number:
Molecular Weight:
227.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

38-42 °C

application(s)

peptide synthesis

functional group

aldehyde

SMILES string

[H]C(=O)CC1CCN(CC1)C(=O)OC(C)(C)C

InChI

1S/C12H21NO3/c1-12(2,3)16-11(15)13-7-4-10(5-8-13)6-9-14/h9-10H,4-8H2,1-3H3

InChI key

PSRHRFNKESVOEL-UHFFFAOYSA-N

Application

Building block employed in a synthesis of (S)-quinuclidine-2-carboxylic acid.[1] Substrate used in an enanioselective organo-catalytic α-vinylation reaction.[2]
Reactant for synthesis of:
  • Pim-1 inhibitors
  • Selective GPR119 agonists for type II diabetes


Reactant for:
  • α-arylation of aldehydes
  • Enantioselective α-benzylation of aldehydes via photoredox organocatalysis
  • Enantioselective α−triflouromethylation of aldehydes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective organo-SOMO catalysis: the alpha-vinylation of aldehydes.
Hahn Kim et al.
Journal of the American Chemical Society, 130(2), 398-399 (2007-12-22)
Tetrahedron Letters, 47, 2515-2515 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service