Skip to Content
Merck
All Photos(1)

Documents

679321

Sigma-Aldrich

n-Propyltriethoxysilane

≥98%, deposition grade

Synonym(s):

Triethoxypropylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3SiCH2CH2CH3
CAS Number:
Molecular Weight:
206.35
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

≥98%

form

liquid

refractive index

n20/D 1.396 (lit.)

bp

179-180 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

CCC[Si](OCC)(OCC)OCC

InChI

1S/C9H22O3Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-9H2,1-4H3

InChI key

NBXZNTLFQLUFES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

137.3 °F - closed cup

Flash Point(C)

58.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that
Céline Hubert et al.
Faraday discussions, 181, 139-146 (2015-04-30)
Silica particles with a controlled number of entropic patches, i.e. dimples, are synthesized through the growth of the silica core of binary multipods that have been produced by a seeded-growth emulsion polymerization reaction. Transmission electron microscopy studies indicate that the

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service