Skip to Content
Merck
All Photos(2)

Documents

678821

Sigma-Aldrich

4-(1-Pyrrolidino)benzaldehyde

97%

Synonym(s):

1-(4-Formylphenyl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

81-85 °C

SMILES string

[H]C(=O)c1ccc(cc1)N2CCCC2

InChI

1S/C11H13NO/c13-9-10-3-5-11(6-4-10)12-7-1-2-8-12/h3-6,9H,1-2,7-8H2

InChI key

DATXHLPRESKQJK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siyang Ding et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-22)
Fluorescent dyes, especially those emitting in the long wavelength region, are excellent candidates in the area of bioassay and bioimaging. In this work, we report a series of simple organic fluorescent dyes consisting of electron-donating aniline groups and electron-withdrawing barbituric
Two-cationic 2-methylbenzothiazole derivatives as green light absorbed sensitizers in initiation of free radical polymerization.
Janina Kabatc et al.
Colloid and polymer science, 293(7), 1865-1876 (2015-07-07)
Elitsa Pavlova et al.
European biophysics journal : EBJ, 44(8), 623-634 (2015-08-01)
The overproduction of free radicals and reactive oxygen species (ROS) has been proved as a basic damage mechanism and cause for oxidative stress. Their measurement is often hindered by the low signal. This could be resolved with the application of

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service