Skip to Content
Merck
All Photos(1)

Key Documents

678570

Sigma-Aldrich

5-Methylfuran-2-carbonyl chloride

97%

Synonym(s):

5-Methyl-2-furoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$151.00

$151.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$151.00

About This Item

Empirical Formula (Hill Notation):
C6H5ClO2
CAS Number:
Molecular Weight:
144.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$151.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

27-31 °C

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

Cc1ccc(o1)C(Cl)=O

InChI

1S/C6H5ClO2/c1-4-2-3-5(9-4)6(7)8/h2-3H,1H3

InChI key

ZURUVZFDEVKNCE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

196.0 °F - closed cup

Flash Point(C)

91.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service