Skip to Content
Merck
All Photos(1)

Key Documents

677078

Sigma-Aldrich

(2S,5S)-(–)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane

kanata purity

Synonym(s):

S,S-Et-RajPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25O2P
CAS Number:
Molecular Weight:
292.35
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]22/D -36°, c = 1 in chloroform

refractive index

n20/D 1.5581

density

1.0760 g/mL at 25 °C

functional group

ether
phosphine

SMILES string

CC[C@H]1CC[C@@H](P1C2=C(C=CC=C2)C3OCCO3)CC

InChI

1S/C17H25O2P/c1-3-13-9-10-14(4-2)20(13)16-8-6-5-7-15(16)17-18-11-12-19-17/h5-8,13-14,17H,3-4,9-12H2,1-2H3/t13-,14-/m0/s1

InChI key

UKBFKBMQNUOKTD-KBPBESRZSA-N

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.
RajPhos is a trademark of Kanata Chemical Technologies, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service