Skip to Content
Merck
All Photos(2)

Key Documents

676179

Sigma-Aldrich

Di-tert-butylneopentylphosphonium tetrafluoroborate

95%

Synonym(s):

Di-tert-butylneopentylphosphine tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H30BF4P
CAS Number:
Molecular Weight:
304.16
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

258-262 °C

functional group

phosphine

SMILES string

F[B-](F)(F)F.CC(C)(C)C[PH+](C(C)(C)C)C(C)(C)C

InChI

1S/C13H29P.BF4/c1-11(2,3)10-14(12(4,5)6)13(7,8)9;2-1(3,4)5/h10H2,1-9H3;/q;-1/p+1

InChI key

UWNPUGYQGIWNRI-UHFFFAOYSA-O

Application

  • Cocatalyst for palladium-catalyzed Kumada couplings
This neopentylphosphine ligand proved to be superior to tris(tert-butyl)phosphine in palladium-catalyzed aminations of aryl bromides and chlorides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lensey L Hill et al.
The Journal of organic chemistry, 71(14), 5117-5125 (2006-07-01)
Di(tert-butyl)neopentylphosphine (DTBNpP) in combination with palladium sources provided catalysts with comparable or better activity for the Hartwig-Buchwald amination of aryl bromides than tri(tert-butyl)phosphine (TTBP) under mild conditions. DTBNpP also provided effective catalysts for amination reactions of aryl chlorides at elevated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service