Skip to Content
Merck
All Photos(1)

Key Documents

674524

Sigma-Aldrich

2-Bromoallyl alcohol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$81.20
10 G
$515.00

$81.20


Estimated to ship on22 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
$81.20
10 G
$515.00

About This Item

Empirical Formula (Hill Notation):
C3H5BrO
CAS Number:
Molecular Weight:
136.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$81.20


Estimated to ship on22 April 2025


Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.5037

bp

58-62 °C/100 mbar

density

1.654 g/mL at 25 °C

functional group

bromo
hydroxyl

SMILES string

OCC(Br)=C

InChI

1S/C3H5BrO/c1-3(4)2-5/h5H,1-2H2

InChI key

MDFFZNIQPLKQSG-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.1 °F - closed cup

Flash Point(C)

76.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service